Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Manganese(II) nitrate hydrate, 99.98% (metals basis), Alfa Aesar™

Manufacturer:  Alfa Aesar 8784822

Quantity 100g

 View more versions of this product

Catalog No. AA8784822


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

15710-66-4
99.98% (metals basis)
178.95
Manganese(II) nitrate hydrate
Lump
Mn(NO3)2·xH2O
MFCD00149788

Specifications

15710-66-4
99.98% (metals basis)
178.95
Odorless
100g
Lump
Mn(NO3)2·xH2O
MFCD00149788
Pink
Manganese(II) nitrate hydrate

Safety and Handling

SDS & Safety Documents

H272-H301-H332-H315-H319-H335: May intensify fire; oxidizer. Toxic if swallowed. Harmful if inhaled. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.

DOTInformation : DOT Class: 5.1, Packaging Group: III

EINECSNumber : 233-828-8

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit