Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Manganese(II) nitrate hydrate, Puratronic™, 99.995% (metals basis), Thermo Scientific Chemicals
Click to view available options
Quantity:
2 g
10 g
50 g
Description
Raw material for electrochemical synthesis of a LaMnO3 thin film coating on stainless steel substrates. Also useful precursors to the oxides of manganese.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | ∽26°C |
Color | Pink |
Physical Form | Crystalline Aggregates |
Quantity | 2 g |
Assay Percent Range | 99.999% (Metal basis) |
Solubility Information | Soluble in water. |
IUPAC Name | manganese(2+);dinitrate;hydrate |
Formula Weight | 178.95 (Anhydrous) |
Odor | Odorless |
Sensitivity | Hygroscopic |
Show More |
Safety and Handling
Hazard Category | H272-H302-H314-H373 |
Precautionary Statement | P210-P220-P221-P260-P264b-P270-P271-P280-P284-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c |
DOTInformation | DOT Class: 5.1, Packaging Group: III |
EINECSNumber | 233-828-8 |
TSCA | Yes |
Recommended Storage | Ambient temperatures; Store under Argon |
RUO – Research Use Only
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction