Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Manganese(II) oxide, 99.99%, (trace metal basis), ACROS Organics™

Click to view available options
Quantity:
1g
10g
Packaging:
Glass bottle
1344-43-0
MnO
70.94
VASIZKWUTCETSD-UHFFFAOYSA-N
manganese ii oxide,manganosite,manganese monoxide,manganese green,cassel green,manganese oxide,rosensthiel green,nu-manese,manganese monooxide,manganese protoxide
14940
oxomanganese
O=[Mn]
1344-43-0
MnO
70.94
VASIZKWUTCETSD-UHFFFAOYSA-N
manganese ii oxide,manganosite,manganese monoxide,manganese green,cassel green,manganese oxide,rosensthiel green,nu-manese,manganese monooxide,manganese protoxide
14940
oxomanganese
O=[Mn]
For ultra trace analysis
Assay Percent Range 76.7 to 78.2% (Mn) (Complexometry)
Solubility Information Solubility in water: insoluble
Formula Weight 70.94
Physical Form Crystalline Powder and Chunks
Percent Purity 99.99%
Grade Trace Metal Basis
Packaging Glass bottle
Solubility Solubility in water: insoluble.
Total Trace Metal Impurities 100ppm max.
Color Green
Melting Point 1650.0°C
Quantity 10g
Chemical Name or Material Manganese(II) oxide
Show More Show Less

GHS H Statement
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiou

GHS Signal Word: Warning

EINECSNumber : 215-695-8