Alfa Aesar™ Manganese(II) sulfate monohydrate, 97%

Manufacturer: Alfa Aesar™ A1180936

Quantity 500g

 View more versions of this product

Catalog No. AAA1180936

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 10034-96-5
    Molecular Formula MnSO4·H2O
    Formula Weight 169.02 (151.00 Anhydrous)
    MDL Number MFCD00149159
    UN Number UN3077
    Merck Index 14,5739
    Chemical Name or Material Manganese (II) sulfate monohydrate

    Specifications

    Formula Weight 169.02 (151.00 Anhydrous)
    Health Hazard 3 GHS P Statement
    P260-P314-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    Get medical advice/attention if you feel unwell.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    DOT Information Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
    Odor Odorless
    Molecular Formula MnSO4·H2O
    MDL Number MFCD00149159
    CAS 10034-96-5
    UN Number UN3077
    Health Hazard 1 Warning
    Health Hazard 2 GHS H Statement
    H373
    May cause damage to organs through prolonged or repeated exposure.
    TSCA TSCA
    Merck Index 14,5739
    pH 3 to 3.5
    Quantity 500g
    RTECS Number OP0893500
    Chemical Name or Material Manganese (II) sulfate monohydrate
    Assay 97%

    Description

    Applications
    Manganese(II) sulfate is used as a colorant in dyes. It is also used as feed additives, catalyst for the synthesis of chlorophyll, make plants grow well and increase production. It is used for making dryer of ink and paint. It is used in paper making , porcelain/ceramic, medicine and other industries.

    Solubility
    Soluble in water. 5-10 g/100 mL at 21°C

    Notes
    Loses all water at 400-450°

    GHS H Statement
    H373
    May cause damage to organs through prolonged or repeated exposure.

    GHS P Statement
    P260-P314-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    Get medical advice/attention if you feel unwell.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit