Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Manganese(II) sulfate monohydrate, 99+%, extra pure, ACROS Organics™

Click to view available options
Quantity:
500g
1kg
5kg
Packaging:
Plastic bottle
10034-96-5
MnO4S·H2O
169.02
MFCD00149159
ISPYRSDWRDQNSW-UHFFFAOYSA-L
manganese sulfate monohydrate, manganese ii sulfate monohydrate, manganese sulfate hydrate, manganese ii sulfate hydrate, mnso4.h2o, manganous sulfate monohydrate, unii-w00lys4t26, manganese 2+ sulfate monohydrate, manganese sulfate usp, manganese 2+ sulfate hydrate
177577
CHEBI:86364
manganese(2+);sulfate;hydrate
O.[O-]S(=O)(=O)[O-].[Mn+2]
10034-96-5
MnO4S·H2O
169.02
MFCD00149159
ISPYRSDWRDQNSW-UHFFFAOYSA-L
manganese sulfate monohydrate, manganese ii sulfate monohydrate, manganese sulfate hydrate, manganese ii sulfate hydrate, mnso4.h2o, manganous sulfate monohydrate, unii-w00lys4t26, manganese 2+ sulfate monohydrate, manganese sulfate usp, manganese 2+ sulfate hydrate
177577
CHEBI:86364
manganese(2+);sulfate;hydrate
O.[O-]S(=O)(=O)[O-].[Mn+2]
Assay Percent Range 99% min. (Complexometry)
Linear Formula MnSO4·H2O
Merck Index 15, 5804
Solubility Information Solubility in water: soluble. Other solubilities: insoluble in alcohol
Formula Weight 169.02
Physical Form Powder
Percent Purity 99+%
Grade Extra Pure
Calcium (Ca) 200ppm max.
Chloride 100ppm max.
Iron (Fe) 20ppm max.
Loss on Ignition 10.0% to 12.0% (500°C)
Magnesium (Mg) 200ppm max.
Packaging Plastic bottle
Solubility Solubility in water: soluble. Other solubilities: insoluble in alcohol
Substances Reducing Permanganate Passes Test
Zinc (Zn) 50ppm max.
Boiling Point 850.0°C
Color Pink to White
Melting Point 700.0°C
Quantity 5kg
Chemical Name or Material Manganese(II) sulfate monohydrate
Show More Show Less

GHS H Statement
May cause damage to organs through prolonged or repeated exposure if inhaled.
Causes serious eye damage.
Toxic to aquatic life with long lasting effects.

GHS P Statement
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Wear protective gloves/protective clothing/eye protection/face protection.
Immediately

GHS Signal Word: Danger