Manganese(II) sulfide, 99.9% (metals basis), Alfa Aesar™

Manufacturer: Alfa Aesar 1283506

Quantity 5g

 View more versions of this product

Catalog No. AA1283506

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 18820-29-6
    Physical Form Powder
    Assay Percent Range 99.9% (Metal basis)
    Molecular Formula MnS
    Formula Weight 87
    MDL Number MFCD00014210
    Chemical Name or Material Manganese(II) sulfide

    Specifications

    Formula Weight 87
    Quantity 5g
    Color Green
    Density 3.99g/cm3 at 20°C
    EINECS Number 242-599-3
    Physical Form Powder
    CAS 18820-29-6
    Assay Percent Range 99.9% (Metal basis)
    Molecular Formula MnS
    Appearance Green powder
    MDL Number MFCD00014210
    Health Hazard 1 H301: Toxic if swallowed.
    Chemical Name or Material Manganese(II) sulfide

    H301: Toxic if swallowed.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit