Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Manganese(II) tungsten oxide, 99.9% (metals basis), Thermo Scientific Chemicals

Catalog Number p-7050738
Click to view available options
Quantity:
50 g
250 g
14177-46-9
MFCD00014212
Manganese tungstate
14177-46-9
MFCD00014212
Manganese tungstate

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Physical Form Powder
Quantity 50 g
Solubility Information Insoluble in water.
Formula Weight 302.79
Percent Purity 99.9%
Odor Odorless
Density 7.23
Assay (metals basis)
Chemical Name or Material Manganese(II) tungsten oxide
Hazard Category H302+H332-H319-H335
Hazard Statement GHS H Statement
H301-H332-H319-H335
Toxic if swallowed.
Harmful if inhaled.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280i-P301+P312-P304+P340-P305+P351+P338-P312-P330-P501c
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit