Alfa Aesar™ Manganese(III) 2,4-pentanedionate

Manufacturer: Alfa Aesar™ 1253018

Quantity 50g

 View more versions of this product

Catalog No. AA1253018

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 14284-89-0
    Formula Weight 352.24
    MDL Number MFCD00000023
    Synonym Manganese(III) acetylacetonate
    Chemical Name or Material Manganese(III) 2,4-pentanedionate

    Specifications

    Formula Weight 352.24
    MDL Number MFCD00000023
    Sensitivity Hygroscopic
    Melting Point ≈160°C (dec.)
    Physical Form Powder
    CAS 14284-89-0
    Synonym Manganese(III) acetylacetonate
    TSCA TSCA
    Solubility Information Soluble in benzene, ethyl acetate
    Quantity 50g
    Chemical Name or Material Manganese(III) 2,4-pentanedionate

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit