Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Manganese(IV) oxide, 88%, electrolytically precipitated, active, ACROS Organics™

Click to view available options
Quantity:
100g
500g
2.5kg
Packaging:
Glass bottle
1313-13-9
MnO2
86.94
MFCD00003463
NUJOXMJBOLGQSY-UHFFFAOYSA-N
manganese dioxide,manganese iv oxide,manganese peroxide,manganese superoxide,manganese black,manganese binoxide,manganese oxide,mangandioxid,black manganese oxide,braunstein
14801
dioxomanganese
O=[Mn]=O
1313-13-9
MnO2
86.94
MFCD00003463
NUJOXMJBOLGQSY-UHFFFAOYSA-N
manganese dioxide,manganese iv oxide,manganese peroxide,manganese superoxide,manganese black,manganese binoxide,manganese oxide,mangandioxid,black manganese oxide,braunstein
14801
dioxomanganese
O=[Mn]=O
Oxidation reagent
Additional Information Oxidation activity:>60% (yield)
Assay Percent Range 83% min. (on dried substance) (Iodometry)
Linear Formula MnO2
Fieser 01,637; 02,257; 03,191; 04,317; 05,422; 06,357; 08,312; 13,233; 14,200; 15,197; 16,202
Merck Index 14,573
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in hydrochloric acid
Formula Weight 86.94
Physical Form Powder
Percent Purity 88%
Density 5.02
Packaging Glass bottle
Solubility Solubility in water: insoluble. Other solubilities: soluble in hydrochloric acid
Color Black
Melting Point 535.0°C
Quantity 100g
Chemical Name or Material Manganese(IV) oxide, 88%
Show More Show Less

GHS H Statement
May intensify fire; oxidizer.
Harmful if swallowed.
Harmful in contact with skin.
May cause damage to organs through prolonged or repeated exposure if inhaled.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Take any precaution to avoid mixing with combustibles.
In case of major

GHS Signal Word: Danger

EINECSNumber : 215-202-6

RTECSNumber : OP0350000

TSCA : TSCA