Learn More
Manganese(IV) oxide, 97%, Thermo Scientific Chemicals
Description
Manganese(IV) oxide is used as a pigment and as a reagent inorganic synthesis especially for the oxidation of allylic alcohols. It is also used for dry -cell batteries such as the alkaline battery and the zinc-carbon battery. It is involved in the preparation of other manganese compounds. It is a precursor to ferromanganese and related alloys, which is useful in the steel industry. It catalyses the decomposition of hydrogen peroxide into oxygen and water. Further, it is used to prepare manganese(II) sulfate, manganese(II) chloride and potassium permanganate. In addition to this, it is used in ceramics and glass manufacturing as an inorganic pigment.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Manganese(IV) oxide is used as a pigment and as a reagent inorganic synthesis especially for the oxidation of allylic alcohols. It is also used for dry -cell batteries such as the alkaline battery and the zinc-carbon battery. It is involved in the preparation of other manganese compounds. It is a precursor to ferromanganese and related alloys, which is useful in the steel industry. It catalyses the decomposition of hydrogen peroxide into oxygen and water. Further, it is used to prepare manganese(II) sulfate, manganese(II) chloride and potassium permanganate. In addition to this, it is used in ceramics and glass manufacturing as an inorganic pigment.
Solubility
Insoluble in water.
Notes
Incompatible with strong acids, strong reducing agents and organic materials.
Specifications
Melting Point | 535°C (decomposition) |
Quantity | 500 g |
Merck Index | 14,5730 |
Solubility Information | Insoluble in water. |
Formula Weight | 86.94 |
Percent Purity | 98% |
Density | 5.026 |
Chemical Name or Material | Manganese (IV) oxide |
Safety and Handling
Hazard Category | H302+H332-H373 |
Hazard Statement | GHS H Statement H302-H332 Harmful if swallowed. Harmful if inhaled. |
Precautionary Statement | P260-P264b-P270-P271-P301+P312-P304+P340-P312-P314-P330-P501c |
EINECSNumber | 215-202-6 |
RTECSNumber | OP0350000 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.