Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Manganese pieces, irregular, 99.9% (metals basis), Thermo Scientific Chemicals

Catalog Number AA1023630
Click to view available options
Quantity:
250 g
1 kg
5 kg
7439-96-5
Mn
54.94
MFCD00011111
PWHULOQIROXLJO-UHFFFAOYSA-N
colloidal, mangan, manganese, elemental, cutaval, magnacat, metal alloy, tronamang, fume, mangan polish, compounds
23930
CHEBI:35154
manganese
[Mn]
7439-96-5
Mn
54.94
MFCD00011111
PWHULOQIROXLJO-UHFFFAOYSA-N
colloidal, mangan, manganese, elemental, cutaval, magnacat, metal alloy, tronamang, fume, mangan polish, compounds
23930
CHEBI:35154
manganese
[Mn]

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color Gray
Appearance Gray irregularly shaped pieces
Density 7.3g/cm3 at 20°C
Assay Percent Range 99.9% (Metals basis)
Quantity 250 g
Formula Weight 54.94
Form Broken Plate
Chemical Name or Material Manganese
EINECSNumber 231-105-1
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit