Manganese Powder, <10 micron, 99.6% (Metals basis), Alfa Aesar™

Manufacturer: Alfa Aesar 45098A1

Quantity 1kg

 View more versions of this product

Catalog No. AA45098A1

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 7439-96-5
    Physical Form Powder
    Assay Percent Range 99.6% (Metals basis)
    MDL Number MFCD00011111
    Chemical Name or Material Manganese

    Specifications

    Quantity 1kg
    Color Gray
    DOT Information DOT Class: 4.1, Packaging Group: III
    Density 7.3g/cm3 at 20°C
    EINECS Number 231-105-1
    Physical Form Powder
    Assay Percent Range 99.6% (Metals basis)
    MDL Number MFCD00011111
    CAS 7439-96-5
    Health Hazard 1 H228: Flammable solid.
    Appearance Gray powder
    Health Hazard 2 Take precautionary measures against static discharges.
    Chemical Name or Material Manganese

    Take precautionary measures against static discharges.

    H228: Flammable solid.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit