Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Manganese Powder, -325 Mesh, 99.95% (Metal Basis), Alfa Aesar™

Click to view available options
Quantity:
25g
100g
7439-96-5
Mn
54.938
MFCD00011111
PWHULOQIROXLJO-UHFFFAOYSA-N
colloidal, mangan, manganese, elemental, cutaval, magnacat, metal alloy, tronamang, fume, mangan polish, compounds
23930
CHEBI:35154
manganese
[Mn]
7439-96-5
Mn
54.938
MFCD00011111
PWHULOQIROXLJO-UHFFFAOYSA-N
colloidal, mangan, manganese, elemental, cutaval, magnacat, metal alloy, tronamang, fume, mangan polish, compounds
23930
CHEBI:35154
manganese
[Mn]
Quantity 25g
Density 7.3g/cm3 at 20°C
Assay Percent Range 99.95%, metal basis
Mesh Size -325 Mesh
Physical Form Powder
Chemical Name or Material Manganese

Take precautionary measures against static discharges.

P210-P240-P241-P264b-P280-P305+P351+P338-P370+P378q

H228-H319

DOTInformation : DOT Class: 4.1; Packaging Group: III

EINECSNumber : 231-105-1

TSCA : Yes

Recommended Storage : Ambient temperatures