Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Manganese Sulfate Monohydrate (Granular Powder/USP/FCC), Fisher Chemical™

Click to view available options
Quantity:
500g
2.5kg
12kg
Packaging:
Poly Bottle
Poly Pail
10034-96-5
H2MnO5S
169.01
MFCD00149159
ISPYRSDWRDQNSW-UHFFFAOYSA-L
manganese sulfate monohydrate, manganese ii sulfate monohydrate, manganese sulfate hydrate, manganese ii sulfate hydrate, mnso4.h2o, manganous sulfate monohydrate, unii-w00lys4t26, manganese 2+ sulfate monohydrate, manganese sulfate usp, manganese 2+ sulfate hydrate
177577
CHEBI:86364
manganese(2+) hydrate sulfate
O.[Mn++].[O-]S([O-])(=O)=O
10034-96-5
H2MnO5S
169.01
MFCD00149159
ISPYRSDWRDQNSW-UHFFFAOYSA-L
manganese sulfate monohydrate, manganese ii sulfate monohydrate, manganese sulfate hydrate, manganese ii sulfate hydrate, mnso4.h2o, manganous sulfate monohydrate, unii-w00lys4t26, manganese 2+ sulfate monohydrate, manganese sulfate usp, manganese 2+ sulfate hydrate
177577
CHEBI:86364
manganese(2+) hydrate sulfate
O.[Mn++].[O-]S([O-])(=O)=O
Arsenic (As) 3mg/kg max. (FCC)
Formula Weight 169.02
Physical Form Solid
Percent Purity 98.0 to 102.0% (USP)
Grade FCC/USP
Density 3.25g/cm³
Identification Pass Test (FCC), Pass Test (USP), Pass Test (FCC/USP)
Lead (Pb) 4mg/kg max. (FCC)
Loss on Ignition 10.0 to 13.0% (USP)
Loss on Heating 10.0 to 13.0% (FCC)
Packaging Poly Bottle
Residual Solvents Meets Requirements (USP)
Selenium (Se) 0.003% max. (FCC)
Substances not ppt by (NH4)2S 0.5% max. (USP)
Decomposition Information 850°C
Boiling Point 850°C
Color Red
Melting Point 700°C
pH 3 to 3.5
Quantity 500g
Chemical Name or Material Manganese Sulfate Monohydrate
Show More Show Less

WARNING!

Emergency Overview
May be harmful if swallowed or inhaled. May cause eye, skin, and respiratory tract irritation. Danger of serious damage to health by prolonged exposure. Toxic to aquatic organisms, may cause long-term adverse effects in the aquatic environment. Hygroscopic. Use personal protective equipment. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. Move to fresh air. If breathing is difficult, give oxygen.

NFPA

Health:1
Flammability:0
Instability:0