Manganous Sulfate, MnSO4-4H2O, MP Biomedicals

$34.69

Chemical Identifiers

Chemical Name or Material Manganous Sulfate
CAS [10034-96-5]
Formula Weight 223.06

  • Products 1
  • Description & Specifications
Products
Catalog Number Mfr. No. Description Price Quantity Availability  

ICN225099

MP Biomedicals
0522509980
100g
Each for $34.69
Check Availability
Add to Cart
Descriptions & Specifications
Safety and Handling

Related Products