Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Manganous Sulfate, Monohydrate, Powder, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™

Click to view available options
Quantity:
500g
2.5kg
Packaging:
Poly Bottle
10034-96-5
H2MnO5S
169.009
ISPYRSDWRDQNSW-UHFFFAOYSA-L
manganese sulfate monohydrate,manganese ii sulfate monohydrate,manganese sulfate hydrate,manganese ii sulfate hydrate,mnso4.h2o,manganous sulfate monohydrate,unii-w00lys4t26,manganese 2+ sulfate monohydrate,manganese sulfate usp,manganese 2+ sulfate hydrate
177577
CHEBI:86364
manganese(2+);sulfate;hydrate
O.[O-]S(=O)(=O)[O-].[Mn+2]
10034-96-5
H2MnO5S
169.009
ISPYRSDWRDQNSW-UHFFFAOYSA-L
manganese sulfate monohydrate,manganese ii sulfate monohydrate,manganese sulfate hydrate,manganese ii sulfate hydrate,mnso4.h2o,manganous sulfate monohydrate,unii-w00lys4t26,manganese 2+ sulfate monohydrate,manganese sulfate usp,manganese 2+ sulfate hydrate
177577
CHEBI:86364
manganese(2+);sulfate;hydrate
O.[O-]S(=O)(=O)[O-].[Mn+2]

High quality chemicals for laboratory and specialized industrial use

Formula Weight 169.02
Physical Form Powder
Grade BAKER ANALYZED™ ACS Reagent
Packaging Poly Bottle
Specific Gravity 1L = 2.95kg
Quantity 500g
Chemical Name or Material Manganous Sulfate, Monohydrate