Learn More
Selleck Chemical LLCSupplier Diversity Partner Manidipine 100mg 89226-50-6  DFS Item
Manufacturer:  Selleck Chemical LLCSupplier Diversity Partner S2481100MG
Manidipine is a calcium channel blocker (dihydropyridine type) that is used clinically as an antihypertensive.
Catalog No. 50-647-4