Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mannitol, Powder, Multi-Compendial, U.S.P., J.T.Baker™

Click to view available options
Quantity:
2.5kg
500g
Packaging:
Poly Bottle

Chemical Identifiers

69-65-8
C6H14O6
182.172
FBPFZTCFMRRESA-KVTDHHQDSA-N
6251
CHEBI:16899
Specific Gravity 1L = 1.52kg
Quantity 2.5kg
CAS 69-65-8
Molecular Formula C6H14O6
InChI Key FBPFZTCFMRRESA-KVTDHHQDSA-N
SMILES C(C(C(C(C(CO)O)O)O)O)O
IUPAC Name (2R,3R,4R,5R)-hexane-1,2,3,4,5,6-hexol
Molecular Weight (g/mol) 182.172
PubChem CID 6251
ChEBI CHEBI:16899
Packaging Poly Bottle
Formula Weight 182.17
Physical Form Powder
Chemical Name or Material Mannitol
Grade Multi-Compendial/USP
Show More Show Less