Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mannose Phosphate Isomerase Mouse anti-Human, Rat, PerCP, Clone: OTI1C7, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP272582PCP
Description
Mannose Phosphate Isomerase Monoclonal specifically detects Mannose Phosphate Isomerase in Human, Rat samples. It is validated for Flow Cytometry.Specifications
Mannose Phosphate Isomerase | |
Monoclonal | |
PerCP | |
PBS with 0.05% Sodium Azide | |
MPI | |
Full length recombinant protein of human Mannose Phosphate Isomerase (NP_002426) produced in HEK293T cells. | |
0.1 mL | |
Primary | |
Human, Rat | |
IgG1 |
Flow Cytometry | |
OTI1C7 | |
Flow Cytometry | |
CDG1B, EC 5.3.1, EC 5.3.1.8, FLJ39201, mannose phosphate isomerase, mannose-6- phosphate isomerase, mannose-6-phosphate isomerase, Phosphohexomutase, Phosphomannose isomerase, phosphomannose isomerase 1, PMI, PMI1phosphohexomutase | |
Mouse | |
Affinity Purified | |
Lipid and Metabolism | |
4351.0 | |
Store at 4°C in the dark. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title