Thermo Scientific™ Manual Crimpers, Decrimpers and Decrimping Pliers Available on GSA/VA Contract for Federal Government customers only.
Achieve secure, repeatable crimps or remove seals with minimized risk of damage.
Manufacturer: Thermo Scientific™ C4032100
 View more versions of this product
Catalog No. 03-375-75B
  Check Availability
Add to Cart
Disclaimers:
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
    Specifications
    For Use With 32mm crimp seals
    Description Crimper, For 32mm crimp seals
    Type Manual Crimper
    Item Description C4032-100
    Vial Closure Size 32mm
    Vial Brand National
    Achieve secure, repeatable crimps or remove seals with minimized risk of damage when you use Thermo Scientific™ Manual Crimpers, Decrimpers and Decrimping Pliers, also for use with National or Chromacol vials.
    Lightweight, precision crimpers attach seals to chromatography or serum vials.
    • Factory set to propery crimp standard-thickness seals
    • Adjustable handle setting scree allows proper crimps with septa of varying thickness
  • Description & Specifications
Related Products
Product Content Feedback