Learn More
Argos Technologies Manual Pipettes
Pinch and release valve system allows for single-hand operation.
Manufacturer:  Argos Technologies P6500V
 View more versions of this product
Catalog No. 03-391-468
Add to cart
Description
Description
  • For aspiration and delivery of lab liquids
  • Pipetting bulbs manufactured from natural rubber
  • For use with serological pipettes up to 50mL
  • Each valve has chemically resistant stainless-steel ball bearing
  • For use with
Specifications
Specifications
serological pipettes up to 50mL
Natural rubber
Purple
Manual pipette