Learn More
Argos Technologies™ Manual Pipettes
Pinch and release valve system allows for single-hand operation.
$47.33 - $47.78
Specifications
Material Natural rubber
Type Manual pipette
For Use With (Equipment) serological pipettes up to 50mL
View More Specs
Products 4
Catalog Number Color Price Quantity & Availability  
Catalog Number Color Price Quantity & Availability  
03-391-465
View Documents
Black
Each for $47.78
Add to cart
03-391-466
View Documents
Green
Each for $47.78
Add to cart
03-391-467
View Documents
Red
Each for $47.33
Add to cart
03-391-468
View Documents
Purple
Each for $47.33
Add to cart
Description
Description
  • For aspiration and delivery of lab liquids
  • Pipetting bulbs manufactured from natural rubber
  • For use with serological pipettes up to 50mL
  • Each valve has chemically resistant stainless-steel ball bearing
  • For use with
Specifications
Specifications
Natural rubber
Manual pipette
serological pipettes up to 50mL