Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MAOA Mouse anti-Human, Clone: OTI2F10, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA525861

Catalog No. PIMA525861


Add to cart

Description

Description

This gene encodes monoamine oxidase A, an enzyme that degrades amine neurotransmitters, such as dopamine, norepinephrine, and serotonin. The protein localizes to the mitochondrial outer membrane. The gene is adjacent to a related gene on the opposite strand of chromosome X. Mutation in this gene results in monoamine oxidase deficiency, or Brunner syndrome.
Specifications

Specifications

MAOA
Monoclonal
0.8 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
P21397
MAOA, Amine oxidase [flavin-containing] A, Monoamine oxidase type A, MAO-A
Mouse
IgG1
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
4128
Immunohistochemistry (Paraffin), Western Blot
OTI2F10
Unconjugated
MAOA
Liquid
MAOA
Human recombit protein fragment corresponding to amino acids 249-484 of MAOA produced in E.coli
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit