Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Lab Vision™ MAP1B/MAP5 (Microtubule-Associated Protein) Ab-1, Mouse Monoclonal Antibody

Ensure accurate, reproducible results in immunohistochemistry experiments with the Thermo Scientific™ MAP1B/MAP5 (Microtubule-Associated Protein) Ab-1, Mouse Monoclonal Antibody.

Specifications

Antigen MAP1B/MAP5 (Microtubule-Associated Protein) Ab-1
Clone 3G5
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Host Species Murine
View More Specs

Products 1
Catalog Number Mfr. No. Formulation Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Formulation Quantity Price Quantity & Availability  
MS248R1
View Documents
Thermo Scientific™
MS248R1
Ready-to-use for IHC (UVLP) 1mL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
Description

Description

Ensure accurate, reproducible results in immunohistochemistry experiments with the Thermo Scientific™ MAP1B/MAP5 (Microtubule-Associated Protein) Ab-1, Mouse Monoclonal Antibody.

Microtubule-associated protein 1B (MAP1B, also known as MAP5, MAP1.2, MAP1(x), or MAP1X) is different from MAP1A (also known as MAP1 or MAP1.1). Reportedly, MAP1B/MAP5 is an early MAP; it is present at high levels in embryonic and newborn rat brain and declines several-fold upon brain maturation. In several cellular situations, MAP1B/MAP5 is the first neuronal MAP to appear, and it is found in neurites from their very first emergence from the cell body. Expression of MAP1B/MAP5 is induced by nerve growth factor.

Host Species: Mouse

Clone: 3G5

Isotype: IgG1

Species Reactivity: Human, Cow and Rat. Does not react with chicken. Others not known.

Epitope: Not determined

Immunogen: Purified bovine brain MAPs

Molecular Weight: 320kDa doublet

Positive Control: Brain

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting (Not verified)
  • Immunofluorescence

Note:

USA: RUO; Int'l: RUO

Specifications

Specifications

MAP1B/MAP5 (Microtubule-Associated Protein) Ab-1
Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Murine
Cancer and Tumor Biology
Purified bovine brain MAPs
3G5
Monoclonal
RUO
Primary
IgG1
SDS
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit