Learn More
MAP1B Rabbit anti-Human, Mouse, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 216331AP
Catalog No. 50-173-3523
Add to cart
Description
Description
The coenzyme NAD and its derivatives are involved in hundreds of metabolic redox reactions and are utilized in protein ADP-ribosylation, histone deacetylation, and in some Ca(2+) signaling pathways. NMNAT.
Specifications
Specifications
MAP1B
0.49 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
P14873, P46821
MAP 1B, MAP1B, MAP5
Rabbit
IgG
150 μL
-20°C
Primary
17755, 4131
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Unconjugated
MAP1B
Liquid
Map1b
MAP1B Fusion Protein Ag16255
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse