Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MAP1LC3A Mouse anti-Human, Mouse, Rat, Clone: 166AT1234, Abnova™

Mouse monoclonal antibody raised against full length recombinant human MAP1LC3A.

Supplier:  Abnova Corporation MAB12280

Catalog No. 89-934-504

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.


Description

Description

MAP1A and MAP1B are microtubule-associated proteins which mediate the physical interactions between microtubules and components of the cytoskeleton. MAP1A and MAP1B each consist of a heavy chain subunit and multiple light chain subunits. The protein encoded by this gene is one of the light chain subunits and can associate with either MAP1A or MAP1B. Two transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq]

Specifications

Specifications

MAP1LC3A
Monoclonal
Unconjugated
Immunohistochemistry (1:50-1:100) Immunofluorescence (1:200) Western Blot (1:1000) The optimal working dilution should be determined by the end user.
LC3/LC3A/MAP1ALC3/MAP1BLC3
Mouse
Protein G purification
RUO
84557
Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
IgG1 κ
Immunofluorescence, Immunohistochemistry (PFA fixed), Western Blot
166AT1234
Mouse monoclonal antibody raised against full length recombinant human MAP1LC3A.
In PBS (0.09% sodium azide)
MAP1LC3A
Recombinant protein corresponding to full length human MAP1LC3A.
400 μL
Primary
Human, Mouse, Rat
Liquid
Product Suggestions

Product Suggestions

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit