Learn More
MAP1LC3A Rabbit anti-Human, Mouse, Rat, Polyclonal , Abnova™
Rabbit polyclonal antibody raised against partial synthetic protein of human MAP1LC3A.
Supplier: Abnova Corporation PAB29558
Description
MAP1A and MAP1B are microtubule-associated proteins which mediate the physical interactions between microtubules and components of the cytoskeleton. MAP1A and MAP1B each consist of a heavy chain subunit and multiple light chain subunits. The protein encoded by this gene is one of the light chain subunits and can associate with either MAP1A or MAP1B. Two transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq]
Specifications
MAP1LC3A | |
Polyclonal | |
Rabbit polyclonal antibody raised against partial synthetic protein of human MAP1LC3A. | |
In PBS (0.09% sodium azide) | |
MAP1LC3A | |
A synthetic peptide (conjugated with KLH) corresponding to amino acids 1-30 at N-terminus of human MAP1LC3A. | |
400 μL | |
Primary | |
Human, Mouse, Rat | |
Liquid |
Immunofluorescence, Immunohistochemistry (PFA fixed), Western Blot | |
Unconjugated | |
Immunofluorescence (1:200) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1:50-100) Western Blot (1:1000) The optimal working dilution should be determined by the end user. | |
LC3/LC3A/MAP1ALC3/MAP1BLC3 | |
Rabbit | |
Ammonium sulfate precipitation | |
RUO | |
84557 | |
Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
Ig |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.