Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MAP2 Mouse anti-Human, Clone:5B7, Abnova™

Mouse monoclonal antibody raised against partial recombinant human MAP2.

Manufacturer:  Abnova Corporation MAB17875

Catalog No. 89-334-365


Add to cart

Description

Description

This gene encodes a protein that belongs to the microtubule-associated protein family. The proteins of this family are thought to be involved in microtubule assembly, which is an essential step in neurogenesis. The products of similar genes in rat and mouse are neuron-specific cytoskeletal proteins that are enriched in dentrites, implicating a role in determining and stabilizing dentritic shape during neuron development. A number of alternatively spliced variants encoding distinct isforms have been described. [provided by RefSeq]
Specifications

Specifications

MAP2
5B7
microtubule-associated protein 2
In PBS (0.05% sodium azide)
Liquid
Mouse
IgG1
Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Monoclonal
Human
ELISA, Flow Cytometry, Western Blot
Unconjugated
ELISA (1:10000)
Flow Cytometry (1:200-1:400)
Western Blot (1:100-1:500)
The optimal working dilution should be determined by the end user.
MAP2
DKFZp686I2148, MAP2A, MAP2B, MAP2C
Recombinant protein corresponding to amino acids 24-123 of human MAP2.
100μg
Antibody
4133
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit