Learn More
MAP2 Mouse anti-Human, Mouse, Rat, DyLight 550, Clone: 5H11, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP192711R
Catalog No. NBP192711R
Add to cart
Description
Description
MAP2 Monoclonal antibody specifically detects MAP2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry, Immunofluorescence.
Specifications
Specifications
MAP2
5H11
Western Blot, Immunocytochemistry/Immunofluorescence
Purified
MAP2
Full length purified bovine MAP2 protein. Epitope maps to 1057-1507 of the human sequence.
Protein G purified
RUO
Store at 4C in the dark.
Monoclonal
Human, Mouse, Rat
Immunocytochemistry, Immunofluorescence, Western Blot
DyLight 550
50mM Sodium Borate with 0.05% Sodium Azide
DKFZp686I2148, MAP-2, MAP2A, MAP2B, MAP2C, microtubule-associated protein 2
Mouse
IgG2b
0.1 ml
Cellular Markers, Cytoskeleton Markers, MAP Kinase Signaling, Neuronal Stem Cells, Neuroscience, Phospho Specific, Stem Cell Markers
Primary
4133