Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MAP2K1 Monoclonal Antibody (OTI2D4), TrueMAB™, OriGene
SDP

Mouse Monoclonal Antibody

Supplier:  OriGene TA505702

Encompass_Preferred

 View more versions of this product

Catalog No. 50-167-6139


May include imposed supplier surcharges.
Only null left
Add to Cart

Description

Description

MEK1 (mitogen activated protein kinase kinase, ERK kinase 1, microtubule-associated protein 2 kinase) is a dual specificity kinase and catalyses both a threonine and a tyrosine residue on MAP kinases ERK1 and ERK2. Mek1 is activated by phosphorylation of serine 218 and 222 residues by Raf1. It is known to be involved in the signaling during stress activate response, apoptosis and proliferative induction by cytokines. Mutations in MEK1 can lead to cardiofaciocutaneous syndrome.
Specifications

Specifications

MAP2K1
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
P31938, Q01986, Q02750
MAP2K1
Full length human recombit protein of human MAP2K1 produced in HEK293T cell.
100 μL
Primary
Canine, Human, Mouse, Rat
Antibody
IgG2a
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
OTI2D4
Unconjugated
MAP2K1
CFC3, MAPKK1, MEK1, MKK1, PRKMK1
Mouse
Affinity Chromatography
RUO
170851, 26395, 478347, 5604
-20° C, Avoid Freeze/Thaw Cycles
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.