Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MAP2K7, Rabbit anti-Human, Clone: R4F9, Abnova™
Rabbit Monoclonal Antibody
Supplier: Abnova Corporation MAB18965
Specifications
MAP2K7 | |
Monoclonal | |
Unconjugated | |
In PBS, pH 7.4 (50% glycerol, 0.02% sodium azide, 0.1% BSA). | |
Jnkk2/MAPKK7/MKK7/PRKMK7 | |
Rabbit | |
200 μL | |
Primary | |
Human, Rat | |
Antibody | |
IgG1 κ |
Flow Cytometry | |
R4F9 | |
Flow Cytometry (1 ug/mL - 0.001 ug/mL) The optimal working dilution should be determined by the end user. | |
mitogen-activated protein kinase kinase 7 | |
MAP2K7 | |
A synthetic phosphopeptide corresponding to residues surrounding S271/T275 of human MAP2K7. | |
RUO | |
5609 | |
Store at -20°C. | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction