Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MAP3K15 Mouse anti-Human, Mouse, Rat, DyLight 405, Clone: OTI1G10, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP272588V
Description
MAP3K15 Monoclonal specifically detects MAP3K15 in Human, Mouse, Rat samples. It is validated for Western Blot.Specifications
MAP3K15 | |
Monoclonal | |
DyLight 405 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
MAP3K15 | |
Recombinant protein fragment corresponding to amino acids 979-1313 of human MAP3K15 (NP_001001671) produced in E.coli. | |
0.1 mL | |
Primary | |
Human, Mouse, Rat | |
IgG1 |
Western Blot | |
OTI1G10 | |
Western Blot | |
Apoptosis signal-regulating kinase 3, ASK3MEKK 15, bA723P2.3, EC 2.7.11, EC 2.7.11.25, FLJ16518, MAPK/ERK kinase kinase 15, MEK kinase 15, mitogen-activated protein kinase kinase kinase 15 | |
Mouse | |
Affinity Purified | |
Protein Kinase | |
389840.0 | |
Store at 4°C in the dark. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title