Learn More
Aviva Systems Biology Corp MAP3K2 Antibody - N-terminal region (ARP49223_P050) | 100uL
Supplier:  Aviva Systems Biology Corp ARP49223_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against MAP3K2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human MAP3K2. Alternate names: MEKK2, MEKK2BApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat, Zebrafish
Catalog No. 50-014-60045