Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MAP3K2 Rabbit anti-Human, Unconjugated, Clone: FHC-13, Abnova™
Rabbit Monoclonal Antibody.
Supplier: Abnova Corporation MAB20483
Description
MAP3K2 Monoclonal antibody specifically detects MAP3K2 in Human samples. It is validated for IP, ICC, IHC, WB-Ce, IF, Flow Cyt.Specifications
MAP3K2 | |
Monoclonal | |
Unconjugated | |
MAP3K2 | |
Rabbit | |
100 μg | |
10746 | |
Store at -20°C for one year. After reconstitution, at 4°C for one month. It can also be aliquotted and stored frozen at -20°C for a longer time. Avoid repeated freezing and thawing. | |
IgG |
Immunoprecipitation, Immunocytochemistry, Immunohistochemistry, Western Blot, Immunofluorescence, Flow Cytometry | |
FHC-13 | |
mitogen-activated protein kinase kinase kinase 2 | |
MEKK2, MEKK2B | |
A synthetic peptide corresponding to human MAP3K2 | |
Primary | |
Human | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction