Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MAP4, Tocris Bioscience™

Click to view available options
Quantity:
10mg
50mg
157381-42-5
C5H10NO5P-2
195.111
HONKEGXLWUDTCF-UHFFFAOYSA-L
(S)-2-Amino-2-methyl-4-phosphonobutanoic acid
71433541
2-azaniumyl-2-methyl-4-phosphonatobutanoate
CC(CCP(=O)([O-])[O-])(C(=O)[O-])[NH3+]
157381-42-5
C5H10NO5P-2
195.111
HONKEGXLWUDTCF-UHFFFAOYSA-L
(S)-2-Amino-2-methyl-4-phosphonobutanoic acid
71433541
2-azaniumyl-2-methyl-4-phosphonatobutanoate
CC(CCP(=O)([O-])[O-])(C(=O)[O-])[NH3+]

Potent group II mGlu agonist. Also specific group III mGlu antagonist

Selective group III metabotropic glutamate receptor antagonist in some electrophysiological systems. Group II/group III agonist in certain neurochemical systems.
Quantity 50mg
Formula Weight 197.13
Percent Purity >96%
Chemical Name or Material MAP4

Recommended Storage : Store at Room Temperature