Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MAP4, Tocris Bioscience™ Available on GSA/VA Contract for Federal Government customers only.

Click to view available options
Quantity:
10mg
50mg
157381-42-5
C5H10NO5P-2
195.111
HONKEGXLWUDTCF-UHFFFAOYSA-L
(S)-2-Amino-2-methyl-4-phosphonobutanoic acid
71433541
2-azaniumyl-2-methyl-4-phosphonatobutanoate
CC(CCP(=O)([O-])[O-])(C(=O)[O-])[NH3+]
157381-42-5
C5H10NO5P-2
195.111
HONKEGXLWUDTCF-UHFFFAOYSA-L
(S)-2-Amino-2-methyl-4-phosphonobutanoic acid
71433541
2-azaniumyl-2-methyl-4-phosphonatobutanoate
CC(CCP(=O)([O-])[O-])(C(=O)[O-])[NH3+]

Potent group II mGlu agonist. Also specific group III mGlu antagonist

Selective group III metabotropic glutamate receptor antagonist in some electrophysiological systems. Group II/group III agonist in certain neurochemical systems.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Quantity 10mg
Formula Weight 197.13
Percent Purity >96%
Chemical Name or Material MAP4

Recommended Storage : Store at Room Temperature