Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MAP4K2 Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Specifications
Antigen | MAP4K2 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP158852
|
Novus Biologicals
NBP158852 |
100 μL |
Each of 1 for $436.00
|
N/A |
Description
MAP4K2 Polyclonal specifically detects MAP4K2 in Human samples. It is validated for Western Blot.Specifications
MAP4K2 | |
Polyclonal | |
Rabbit | |
Signal Transduction | |
Q12851 | |
5871 | |
Synthetic peptides corresponding to MAP4K2(mitogen-activated protein kinase kinase kinase kinase 2) The peptide sequence was selected from the N terminal of MAP4K2. Peptide sequence HLHPMRALMLMSKSSFQPPKLRDKTRWTQNFHHFLKLALTKNPKKRPTAE. | |
Primary | |
Store at -20C. Avoid freeze-thaw cycles. |
Western Blot | |
Unconjugated | |
RUO | |
PBS and 2% Sucrose with 0.09% Sodium Azide | |
B lymphocyte serine/threonine protein kinase, B lymphocyte serine/threonine-protein kinase, BL44, EC 2.7.11, EC 2.7.11.1, GC kinase, GCKMEK kinase kinase 2, Germinal center kinase, germinal centre kinase (GC kinase), MAPK/ERK kinase kinase kinase 2, mitogen-activated protein kinase kinase kinase kinase 2, Rab8 interacting protein, Rab8-interacting protein, RAB8IPMEKKK 2 | |
MAP4K2 | |
IgG | |
Affinity Purified | |
91 kDa |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title