Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MAP4K3 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA550275
Description
The antibody detects endogenous levels of total MAP4K3 protein.
MAP4K3 Polyclonal antibody specifically detects MAP4K3 in Human, Mouse samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.Specifications
MAP4K3 | |
Polyclonal | |
Unconjugated | |
MAP4K3 | |
4833416M01Rik; 4833416M07Rik; 9530052P13Rik; germinal center kinase-like kinase; germinal center kinase-related protein kinase; GLK; MAP4K3; MAPK/ERK kinase kinase kinase 3; MAPKKKK3; MEK kinase kinase 3; MEKKK 3; MEKKK3; mitogen activated protein kinase kinase kinase kinase 3; mitogen-activated protein kinase kinase kinase kinase 3; RAB8IPL1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
225028, 8491 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1.3 mg/mL | |
PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
Q8IVH8, Q99JP0 | |
MAP4K3 | |
Fusion protein corresponding to a region derived from internal residues of human Mitogen-activated protein kinase kinase kinase kinase 3 | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction