Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MAPA™ TriLites 984 Extended Length Tripolymer Gloves

Catalog Number 19165716
Click to view available options
Size:
Small
Medium
Large
X-Large
This item is not returnable. View return policy
These products contain natural rubber latex, a component of which may cause allergic reaction in some users. Use only as directed and in accordance with manufacturer instructions and warnings. Please consult your institution's policies regarding use of these products.
This item is not returnable. View return policy
These products contain natural rubber latex, a component of which may cause allergic reaction in some users. Use only as directed and in accordance with manufacturer instructions and warnings. Please consult your institution's policies regarding use of these products.

Extra long, 11.5 in. length offers extended protection for the wrist and upper forearm when working with chemicals.

Tripolymer blend of neoprene, nitrile and natural rubber provides comfort of latex exam gloves with added strength, tear resistance and chemical splash protection

  • Exclusive finish minimizes tape and label sticking to gloves
  • Textured grip with lightly halogenated finish for optimum tactility and grip; wet or dry
  • Ambidextrous design, rolled cuff, unlined
  • Powder-free, clean process (CP) for no cross contamination
  • Thickness: 6 mil
  • Length: 11.5 in. (29 cm)
  • Purple

Applications:

Laboratory research, light assembly and sorting, precision parts handling, light maintenance

Certifications/Compliance ASTM D3578, 21 CFR 170-199
Certifications/Compliance (Europe) Cat 3 CE 0334
Chlorinated Yes
Color Purple
Hand Ambidextrous
Latex Free No
Material Tripolymer
Powdered Powder Free
Size Medium
Thickness (English) 6 mil
Thickness (Metric) 0.15 mm
EN ISO 374-1 Type B
EN ISO 374-1 CHEMICAL LETTER CODES KPT
EN ISO 374 - 5 Bacteria, Fungi only
For Use With (Application) Laboratory research, light assembly and sorting, precision parts handling, light maintenance
Glove Cuff Style Rolled
Grip Design Pebbled
Lining Unlined
Model 984927
Series TriLItes 984
Length (English) 11.5 in.
Length (Metric) 29.2 cm
Show More Show Less
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit