MAPA™ AdvanTech™ Rollpruf™ Special Formula Gloves
Exclusive cuff design prevents rolldown
Manufacturer: MAPA™ 513983
 View more versions of this product
Catalog No. 11-394-68
  Check Availability
Add to Cart
    Specifications
    Disposable Yes
    Exterior Finish Textured
    Glove Cuff Style Rolled
    For Use With (Application) semiconductor, electronics, aerospace, pharmaceutical and biotechnology industries, no pigments, fillers or DOP Plasticizers, resist Acids, Salts, Alcohols, Ketones and Alkalis
    Length (English) 12 in.
    Latex Free No
    Length (Metric) 30.5cm
    Material Tri-polymer
    Powdered Yes
    Size 7
    Thickness 8mil
    Color Black
  • Description & Specifications
Related Products
Product Content Feedback