MAPA™ Classics™ L-200 Gloves
Prevents greasy solids build-up
Manufacturer: MAPA™ 158568
 View more versions of this product
Catalog No. 19-999-972D
  Check Availability
Add to Cart
    Specifications
    For Use With (Application) Resistant to mild Acids, Alcohols, Alkalis, Salts, Aldehydes and some animal and vegetable Fats, excellent resistance to abrasions, cuts and tears, food applications, electronic assembly, lab and agricultural chemical handling, Glassware handling and commercial housekeeping.
    Certifications ISO, 21 CFR 170-199, CE
    Exterior Finish Raised diamond
    Glove Cuff Style Flat
    Length (English) 12 in.
    Length (Metric) 30.5cm
    Material Natural Rubber
    Palm Type Contoured
    Powdered No
    Size 8
    Thickness 18mil
    Color Dark blue
  • Description & Specifications
Related Products
Product Content Feedback