MAPA™ Kroflex™ 840 Natural Rubber Gloves

Enhanced non-slip feature offers ultra-protection when handling heavy/sharp objects in wet environments

Manufacturer: MAPA™ 840410

 View more versions of this product

Catalog No. 19-249-753

  Check Availability
Add to Cart

Alert

This product contains natural rubber latex and is not recommended for use by those who may be allergic to natural rubber proteins.


    Specifications

    Glove Cuff Style Knit
    For Use With (Application) handling Glass, hot Plastic parts and waste treatment applications
    Length (English) 10.5 in.
    Length (Metric) 26.5cm
    Material HDPE
    Size 10
    Weight Heavy weight
    Coating Material Rough
    Color Gray, blue
    Cut Resistance Rating Level 4

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit