Learn More
MAPA™ Krynit™ 563 Cut-Resistant Gloves
Provides cut/liquid protection for intricate handling in oily environments
Manufacturer:  MAPA™ 563410
 View more versions of this product
Catalog No. 19-249-774
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
Smooth
Knit
mechanical maintenance, stacking Oily panels, pressing Metal, handling Metal sheets
10
9.85 in.
Level 2
Hand-specific
HDPE (High-density Polyethylene),Polyamide
Black,Gray
25cm