MAPA™ Krynit™ 582 Cut Resistance Gloves

Provides high-performance protection for intricate handling in oily environnement

Manufacturer: MAPA™ 582417

 View more versions of this product

Catalog No. 19-249-800

  Check Availability
Add to Cart

    Specifications

    Glove Cuff Style Knit
    Hand Hand-specific
    For Use With (Application) mechanical maintenance, stacking Oily panels, pressing Metal, handling Metal sheets
    Length (English) 9 in.
    Length (Metric) 22.8cm
    Material HDPE
    Size 7
    Weight Heavy weight
    Coating Material Seamless
    Color Black
    Cut Resistance Rating Level 5

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit