Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MAPA™ Krynit™ 582 Cut Resistance Gloves

Provides high-performance protection for intricate handling in oily environnement

$4.88 - $23.55

Save up to 25%

Specifications

Coating Material Seamless
Color Black
Cut Resistance Rating Level 5
Material HDPE
For Use With (Application) mechanical maintenance, stacking Oily panels, pressing Metal, handling Metal sheets
View More Specs

Products 5
Catalog Number Mfr. No. Length (English) Length (Metric) Size Price Quantity & Availability  
Catalog Number Mfr. No. Length (English) Length (Metric) Size Price Quantity & Availability  
19-249-801
View Documents
MAPA™
582418
9.35 in. 23.7 cm 8 Pair for $23.55
Case of 48 PR for $937.00
Add to cart
 
19-249-802
View Documents
MAPA™
582419
9.85 in. 25.1 cm 9 Pair for $23.55
Case of 48 PR for $937.00
Add to cart
 
19-249-803
View Documents
MAPA™
582410
10.25 in. 26.3 cm 10 Pair for $23.55
Case of 48 PR for $937.00
Add to cart
 
19-249-804
View Documents
MAPA™
582411
10.5 in. 26.6 cm 11 Pair for $4.88

Save 25% Reg : $6.51

Case of 48 PR for $234.00

Save 75% Reg : $937.00

Add to cart
 
19-249-800
View Documents
MAPA™
582417
9 in. 22.8 cm 7 Pair for $23.55
Case of 48 PR for $937.00
Add to cart
 
Description

Description

  • Special Grip&Proof coating from nitrile for better liquid proofness and good grip to oily contact
  • Heavy duty
  • Knitted wrist
  • Seamless textile support from HDPE fibres on the interior wall
  • Non-slip grip
  • Optimal cut resistance: level 5
  • Highly resistant to abrasion
  • A visible indicator in blue textile for easy identification of cut-performance level in the workplace
  • Guaranteed gloves performance even after five cleaning cycles
  • Color: Black
  • Length: 9 to 10.5 in.

  • Maintenance in wet environments
  • Handling glass sheets
  • Mechanical maintenance in automotive/mechanical industry
  • Pressing metal, handling metal sheets
  • Sorting small parts
  • Stacking oily panels

Certifications

CE marked, ISO 13997

Specifications

Specifications

Seamless
Level 5
mechanical maintenance, stacking Oily panels, pressing Metal, handling Metal sheets
Hand-specific
Black
HDPE
Knit
Heavy weight
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit