MAPA™ Medium Weight StanSolv™ Gloves

Manufacturer: MAPA™ 492829

 View more versions of this product

Catalog No. 19-130-2653D

  Check Availability
Add to Cart

    Specifications

    Glove Cuff Style Flat
    For Use With (Application) pesticides, cleaning of printing-press rollers, maintenance, timber treatment and finishing
    Length (English) 12.5 in.
    Length (Metric) 31.7cm
    Lining Cotton Flock
    Material Nitrile
    Size 9
    Thickness 18mil
    Coating Material Embossed, Diamond
    Color Green

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit