MAPA™ Optinit Nitrile Gloves

Manufacturer: MAPA™ 472416

 View more versions of this product

Catalog No. 19-130-2641A

  Check Availability
Add to Cart

    Specifications

    Glove Cuff Style Flat
    For Use With (Application) laboratory and food application where dexterity, plus mild chemical/liquid protection is required
    Length (English) 12 in.
    Length (Metric) 30.5cm
    Material Nitrile
    Powdered No
    Size 6
    Thickness 8mil
    Coating Material Pebble
    Color Blue

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit