MAPA™ Ultrane™ 561 Gloves
Manufacturer: MAPA™ 561416
 View more versions of this product
Catalog No. 19-161-419
  Check Availability
Add to Cart
    Specifications
    Glove Cuff Style Knit
    Length (English) 8 in.
    Length (Metric) 20.32cm
    Lining Polyamide
    Material Nitrile
    Size 6
    Coating Material Rough
    Color Gray, black
  • Description & Specifications
Related Products
Product Content Feedback