MAPA™ Ultrane™ 562 Gloves
Manufacturer: MAPA™ 562418
 View more versions of this product
Catalog No. 19-161-426
  Check Availability
Add to Cart
    Specifications
    Glove Cuff Style Knit
    For Use With (Application) automotive, mechanical industry, sorting small parts, screwing and bolting, pump manufacturing, precision assembly, Metal part assembly
    Length (English) 8.5 in.
    Length (Metric) 21.5cm
    Lining Polyamide
    Material Nitrile
    Size 8
    Coating Material Raised dots
    Color Black, gray
  • Description & Specifications
Related Products
Product Content Feedback