MAPA™ Ultrane™ 562 Gloves
Manufacturer: MAPA™ 562419
 View more versions of this product
Catalog No. 19-161-427
 N/A
N/A
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 
    Specifications
    Glove Cuff Style Knit
    For Use With (Application) automotive, mechanical industry, sorting small parts, screwing and bolting, pump manufacturing, precision assembly, Metal part assembly
    Length (English) 8.5 in.
    Length (Metric) 21.5cm
    Lining Polyamide
    Material Nitrile
    Size 9
    Coating Material Raised dots
    Color Black, gray
  • Description & Specifications
Related Products
Product Content Feedback